Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC27 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5894 Supplier Bioss Supplier No. BS4096R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae protein Cdc27, and the gene product of Schizosaccharomyces pombe nuc 2. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction. This protein was shown to interact with mitotic checkpoint proteins including Mad2, p55CDC and BUBR1, and thus may be involved in controlling the timing of mitosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen CDC27
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDC27
Gene Accession No. P30260
Gene Alias AI452358; AN CDC27Hs; ANAPC3; anaphase promoting complex subunit 3; Anaphase-promoting complex subunit 3; anaphase-promoting complex, protein 3; APC3; BC023187; BOS_19285; CDC27; CDC27 homolog; cdc27.S; CDC27Hs; cell division cycle 27; cell division cycle 27 homolog; cell division cycle 27 homolog (S. cerevisiae); cell division cycle 27 S homeolog; Cell division cycle protein 27 homolog; D0S1430E; D17S978E; HNUC; H-NUC; NUC2; nuc2 homolog; XELAEV_18046079mg
Gene Symbols CDC27
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDC27/ANAPC3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 996
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.