Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDC2L5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599763 Supplier Invitrogen™ Supplier No. PA599763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDK13.

CDK13/cyclin K serine/threonine kinase is dimeric protein complex. CDK13 is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are well known for their essential roles as master switches in cell cycle control. The exact function of this protein has not yet been determined, but it may play a role in mRNA processing and may be involved in regulation of hematopoiesis. Cyclin K is a member of the transcription cyclin family. These cyclins may regulate transcription through their association with and activation of cyclin-dependent kinases (CDK) that phosphorylate the C-terminal domain of the large subunit of RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC2L5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdk13
Gene Accession No. Q14004, Q69ZA1
Gene Alias 2310015O17Rik; CDC2L; Cdc2l5; CDC2-related protein kinase 5; CDK13; cell division cycle 2-like 5 (cholinesterase-related cell division controller); cell division cycle 2-like protein kinase 5; Cell division protein kinase 13; CHED; Cholinesterase-related cell division controller; cyclin dependent kinase 13; cyclin-dependent kinase 13; hCDK13; Kiaa1791
Gene Symbols Cdk13
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDK13(Accession Q14004), corresponding to amino acid residues T670-Y720.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69562, 8621
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.