Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc2L6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599764 Supplier Invitrogen™ Supplier No. PA599764
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CD2L6.

CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc2L6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdk19
Gene Accession No. Q8BWD8, Q9BWU1
Gene Alias 2700084L06Rik; AI845279; AW228747; bA346C16.3; Cdc2l6; CDC2-related protein kinase 6; Cdk11; Cdk19; CDK8-like cyclin-dependent kinase; cell division cycle 2-like 6 (CDK8-like); cell division cycle 2-like protein kinase 6; Cell division protein kinase 19; cyclin dependent kinase 19; cyclin dependent kinase 19 variant 2; cyclin-dependent kinase (CDC2-like) 11; cyclin-dependent kinase 11; Cyclin-dependent kinase 19; death-preventing kinase; KIAA1028; mKIAA1028
Gene Symbols Cdk19
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDK19(Accession Q9BWU1), corresponding to amino acid residues L431-L481.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23097, 78334
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.