Learn More
Invitrogen™ Cdc34 Polyclonal Antibody

Description
Recommended positive controls: 293.
Specifications
Specifications
| Antigen | Cdc34 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with no preservative |
| Gene | CDC34 |
| Gene Accession No. | P49427 |
| Gene Alias | (E3-independent) E2 ubiquitin-conjugating enzyme R1; AI327276; CDC34; cell division cycle 34; cell division cycle 34 homolog; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; serine protease; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme Cdc34; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 |
| Gene Symbols | CDC34 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.