Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC37 Antibody (6B3B5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261731
Description
CDC37 Monoclonal specifically detects CDC37 in Human samples. It is validated for Western Blot, ELISA.Specifications
CDC37 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
Mouse | |
24.8 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
6B3B5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
CDC37 | |
Purified recombinant fragment of human CDC37 (AA: 241-378) expressed in E. Coli. | |
Protein G purified | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific | |
11140 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction