Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC37, p Ser13 Antibody (3B3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CDC37 |
---|---|
Clone | 3B3 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human samples. It is validated for Western Blot, ELISASpecifications
CDC37 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
A synthesized peptide derived from Human CDC37 [p Ser13] [UniProt Q16543] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3B3 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
11140 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title