Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582126 Supplier Invitrogen™ Supplier No. PA582126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGFSKSMVNT KPEKTEEDSE EVREQKHKTF VEKYEKQIKH FGMLRRWDDS QKYLSDNVHL VCEETANYLV IWCIDLEVEE KCALMEQVAH QTIVMQFILE LAKSLKVDPR ACFRQFFTKI KTADRQYMEG FNDELEAFKE RV Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

In S. cerevisiae, Cdc37 has been shown to be an important regulator of cell division cycle through its action on cyclin-dependent kinases (CDK's). Cdc37 has also been associated with signaling pathways and kinases unrelated to cell cycle control, including v-Src, casein kinase II, MPS1 kinase and the sevenless receptor tyrosine kinase. Interestingly, HSP90 has also been shown to be associated with many of the same cellular targets as Cdc37. It has now been shown that Cdc37 is the same protein previously described as p50 which has been found to be associated with many of the same proteins as HSP90. Among others, Cdc37/p50 has been found complexed with Src-family kinases (Fyn, Yes, Fes, Lck & Fgr), Raf and pp60v-src. More recently, Cdc37 has been shown to be a chaperone independently and in concert with HSP90. In fact, Cdc37 is necessary for the binding of HSP90 to Cdk4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc37
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC37
Gene Accession No. Q16543
Gene Alias CDC 37; CDC37; CDC37 (cell division cycle 37 S. cerevisiae homolog); CDC37 (cell division cycle 37, S. cerevisiae, homolog); CDC37 cell division cycle 37 homolog; Cdc37 homolog; CDC37 protein; CDC37A; Cdc37p; Cell division control protein 37; cell division cycle 37; cell division cycle 37 homolog; hsp90 chaperone protein kinase-targeting subunit; Hsp90 co-chaperone Cdc37; Hsp90 co-chaperone Cdc37, N-terminally processed; p50; P50CDC37; SMO1; YD9489.03; YDR168W
Gene Symbols CDC37
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDC37. Recombinant protein control fragment (Product #RP-102063).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.