Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC37 Rabbit anti-Human, Mouse, Rat, Clone: 6X0A3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CDC37 |
---|---|
Clone | 6X0A3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CDC37 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
11140 | |
IgG | |
Affinity purified |
6X0A3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDC37 (Q16543). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title