Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc37L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582748 Supplier Invitrogen™ Supplier No. PA582748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHNSESLDQE HAKAQTAVSE LRQREEEWRQ KEEALVQREK MCLWSTDAIS KDVFNKSFIN QDKRKDTEDE DKSE Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.

CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc37L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC37L1
Gene Accession No. Q7L3B6
Gene Alias 2700033A15Rik; BB159850; CDC37 cell division cycle 37 homolog-like 1; CDC37B; Cdc37l; Cdc37l1; cell division cycle 37 homolog-like 1; cell division cycle 37 like 1; cell division cycle 37-like 1; HARC; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5
Gene Symbols CDC37L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDC37L1. Recombinant protein control fragment (Product #RP-92901).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.