Learn More
Invitrogen™ CDC37L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596304
Description
Immunogen sequence: EQPWPPPGPW SLPRAEGEAE EESDFDVFPS SPRCPQLPGG GAQMYSHGIE LACQKQKEFV KSSVACKWNL AEAQQKLGSL ALHNSESLDQ EHAKAQTAVS ELRQREEEWR QKEEALVQRE KMCLWSTDAI SKDVFNKSFI NQDKRKDTED EDKSESFMQK YEQKIRHFGM LSRWDDSQRF LSDHPYLVCE ETAKYLILWC FHLEAEKKGA LMEQIAHQAV VMQFIMEMAK NCNVDPRGC; Positive Samples: A-549, 293T, HeLa, Mouse kidney; Cellular Location: Cytoplasm.
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008].
Specifications
CDC37L1 | |
Polyclonal | |
Unconjugated | |
CDC37L1 | |
2700033A15Rik; BB159850; CDC37 cell division cycle 37 homolog-like 1; CDC37B; Cdc37l; Cdc37l1; cell division cycle 37 homolog-like 1; cell division cycle 37 like 1; cell division cycle 37-like 1; HARC; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55664, 67072 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.66 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7L3B6, Q9CZP7 | |
CDC37L1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 2-240 of human CDC37L1 (NP_0603832). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.