Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC40 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212781DL594
Description
CDC40 Polyclonal antibody specifically detects CDC40 in Human samples. It is validated for Western BlotSpecifications
CDC40 | |
Polyclonal | |
Western Blot | |
Cell division cycle 40 homolog, cell division cycle 40 homolog (S. cerevisiae), cell division cycle 40 homolog (yeast), Ehb3, EH-binding protein 3, hPRP17, MGC102802, pre-mRNA splicing factor 17, pre-mRNA-processing factor 17, PRP17 homolog, PRP17EHB3PRPF17FLJ10564 | |
The immunogen this antibody was made to, maps to a region between residue 175 and 225 of human Cell Division Cycle 40 using the numbering given in entry NP_056975.1 (GeneID 51362). | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication | |
51362 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction