Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC42 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP176330MFV500
Description
CDC42 Polyclonal antibody specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
CDC42 | |
Polyclonal | |
Western Blot, ELISA | |
CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human CDC42. The immunogen is located within amino acids 40 - 90 of CDC42. Amino Acid Squence: FDTAGQEDYDRLRPLSY | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific, Signal Transduction | |
998 | |
Store at 4C in the dark. | |
IgY |
Western Blot, ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Chicken | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction