Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDC42 Monoclonal Antibody (M152)
GREENER_CHOICE

Catalog No. PIMA547705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547705 Supplier Invitrogen™ Supplier No. MA547705
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 21 kDa* protein in human Jurkat cells and mouse brain. It does not recognize a human RhoA GST fusion protein.

Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M152
Concentration 0.125 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CDC42
Gene Accession No. P60766, P60953, Q8CFN2
Gene Alias AI747189; AU018915; CDC42; CDC42Hs; Cell division control protein 42 homolog; cell division cycle 42; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K; G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; RP1-224A6.5; small GTP binding protein CDC42; TKS
Gene Symbols CDC42
Host Species Mouse
Immunogen Clone (M152) was generated from a rat recombinant Cdc42 protein. This sequence is highly conserved in human and mouse Cdc42.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12540, 64465, 998
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.