Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc42, Mouse anti-Mouse, Yeast, Human, Clone: 28-10, Millipore Sigma™

Catalog No. MABN2485100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2485100 100 μL
MABN248525 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2485100 Supplier MilliporeSigma Supplier No. MABN2485100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cdc42, clone 28-10, Cat. No. MABN2485, is a highly specific mouse monoclonal antibody that targets Cell division control protein 42 (Cdc42) and has been tested for use in Immunocytochemistry and Western Blotting.

Cell division control protein 42 (UniProt: P19073; also known as Cdc42, Suppressor of RHO3 protein 2) is encoded by the CDC42 (also known as SRO2, TLR229C, L8083.13) gene (Gene ID: 850930) in yeast (Saccharomyces cerevisiae). Cdc42 is a member of the Rho GTPase family that regulates multiple signaling pathways involved in actomyosin organization, cell proliferation, and maintenance of cell polarity in most eukaryotic cell types. Cdc42 signaling process involves guanine nucleotide exchange factors (GEFs), guanine nucleotide dissociation inhibitors (GDIs), and GTPase-activating proteins (GAPs). In yeast, generation and maintenance of robust Cdc42 polarity promotes membrane expansion during bud formation and it has been shown that Cdc42 is dynamically maintained at the polarity cap through its continuous cycling between the polarity cap and internal pools. Cdc42 is highly polarized during bud emergence and is specifically required for exocytosis at this stage. Cdc42 is synthesized with a propeptide region (aa 189-191), which is cleaved off in the mature form. The mature form contains three nucleotide (GTP)-binding sites. (Ref.: Wu, H., and Brennwald, P. (2010) Mol. Cell Biol. 30(21); 5207-5217).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 28-10
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Cdc42 in Western Blotting applications (Sullivan, C.S., et. al. (2016). J Biol Chem. 291(51):26262-26272; Wu, H., et. al. (2010). Mol Cell Biol. 30(21):5207-17).Immunocytochemistry Analysis: A representative lot detected Cdc42 in Immunocytochemistry applications (Wu, H., et. al. (2010). Mol Cell Biol. 30(21):5207-17).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Cell division control protein 42;Suppressor of RHO3 protein 2
Gene Symbols CDC42;SRO2;TLR229C;L8083.13
Host Species Mouse
Immunogen GST-tagged full-length recombinant yeast Cdc42.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_013330
Test Specificity Clone 28-10 specifically detects Cell division control protein 42 (Cdc42).
Target Species Mouse, Yeast, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.