Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CDC42, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1646MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1646MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1646MI Supplier MilliporeSigma™ Supplier No. ABN1646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cell division control protein 42 homolog (UniProt: P60953; also known as G25K GTP-binding protein) is encoded by the CDC42 gene (Gene ID 998) in human. CDC42 homolog is a plasma membrane-associated small GTPase that cycles between an active GTP-bound and an inactive GDP-bound state. Its activity is regulated by guanine nucleotide exchange factors (GEFs), which promote the exchange of bound GDP for free GTP, GTPase activating proteins (GAPs) that increase the GTP hydrolysis activity, and GDP dissociation inhibitors, which inhibit the dissociation of the nucleotide from the GTPase. In the active state it binds to a variety of effector proteins to regulate cellular responses. Cdc42 is known to regulate the dynamic organization of the cytoskeleton and membrane trafficking for physiologic processes such as cell proliferation, motility, polarity, cytokinesis and growth. Deregulation of Cdc42 is found in several pathogenic processes such as cancer, cardiovascular diseases, and neuronal degenerative diseases. It is involved in epithelial cell polarization processes and regulates the bipolar attachment of spindle microtubules to kinetochores before chromosome congression in metaphase. During the prometaphase stage, it localizes to spindle and moves to the central spindle as cells progress through anaphase to telophase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS without preservatives.
Gene CDC42
Gene Accession No. P60953-1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal end sequence of mature human CDC42 brain isoform.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_426359
Test Specificity This polyclonal antibody is purified from an antiserum that has been shown to specifically detect recombinant human CDC42 brain isoform Cdc42-palm (UniProt P60953-1) with or without C188S mutation, but not the placenta isoform Cdc42-prenyl (UniProt P60953-2) by Western blotting analysis (Kang, R., et al. (2008). Nature. 456(7224):904-909). Note that the designation of CDC42 isoform 1 and 2 by Kang et al. (2008) is the exact opposite of that by UniProt P60953 (human) and P60766 (mouse), while both Kang et al. (2008) and UniProt Q8CFN2 (rat) designate the rat CDC42 brain isoform as isoform 2.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.