Learn More
CDC42BPB/MRCK beta Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304893AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
Protein kinases comprise a large group of encoded factors that regulate cellular processes by catalyzing the transfer of a phosphate group to a hydroxyl acceptor in serine, threonine or tyrosine residues. Myotonic dystrophy kinaserelated Cdc42-binding (DMPK-like) kinases-alpha and beta (MRCK-alpha, beta) contain a cysteine-rich motif and a putative pleckstrin homology domain. MRCKs can phosphorylate nonmuscle Myosin light chain and influences Actin-Myosin contractility. MRCK-alpha can phosphorylate and activate LIM kinases downstream of Cdc42, which leads to inactivation of ADF/Cofilin and to Actin cytoskeletal reorganization. MRCK-alpha can also influence neurite outgrowth promoted by Cdc42 and Rac.Specifications
CDC42BPB/MRCK beta | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y5S2 | |
CDC42BPB | |
Between 750 and 800 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CDC42BPB | |
Serine/threonine-protein kinase MRCK beta, CDC42-binding protein kinase beta, CDC42BP-beta, DMPK-like beta, Myotonic dystrophy kinase-related CDC42-binding kinase beta, MRCK beta,Myotonic dystrophy protein kinase-like beta, CDC42BPB, KIAA1124 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9578 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.