Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC42EP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CDC42EP2 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7484
![]() ![]() |
Proteintech Group Inc
118241AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Cdc42, a small GTPase, regulates Actin polymerization, elongation of cell shape and cell signaling through interactions with many different downstream effector proteins, most of which contain a Cdc42-binding motif known as a CRIB domain. Cdc42EP2 (Cdc42 effector protein 2), also known as BORG1 or CEP2, is a 210 amino acid intracytoplasmic protein that localizes to both the cytoplasm and the cytoskeleton and contains one CRIB domain. Highly expressed in heart and present at lower levels in liver and pancreas, Cdc42EP2 interacts with Cdc42 and is thought to be involved in the organization of the Actin cytoskeleton and may also influence Actin filament assembly and overall cell shape.Specifications
CDC42EP2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O14613, Q5PQP4, Q8JZX9 | |
104252, 10435, 309175 | |
CDC42EP2 Fusion Protein Ag2392 | |
Primary | |
-20°C | |
CDC42EP2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Binder of Rho GTPases 1, BORG1, Cdc42 effector protein 2, CDC42EP2, CEP2 | |
Cdc42ep2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title