Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc42EP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550002 Supplier Invitrogen™ Supplier No. PA550002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CDC42EP3 / BORG2 protein.

CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. This protein can interact with CDC42, as well as with the ras homolog gene family, member Q (ARHQ/TC10). Expression of this protein in fibroblasts has been shown to induce pseudopodia formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDC42EP3
Gene Accession No. Q9CQC5, Q9UKI2
Gene Alias 3200001F04Rik; AA986861; Binder of Rho GTPases 2; BORG2; CDC42 effector protein (Rho GTPase binding) 3; CDC42 effector protein 3; Cdc42ep3; CEP3; CRIB-containing BORG2 protein; MSE55-related Cdc42-binding protein; MSE55-related protein; UB1
Gene Symbols CDC42EP3
Host Species Rabbit
Immunogen Synthesized peptide derived from N-terminal of human CDC42EP3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10602, 260409, 313838
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.