Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc42EP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584517
Description
Immunogen sequence: DISFLQGNYE LLPGNQEKAH LGQFPGHNEF FRANSTSDSV FTETPSPVLK NA.
CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. This protein can interact with CDC42, as well as with the ras homolog gene family, member Q (ARHQ/TC10). Expression of this protein in fibroblasts has been shown to induce pseudopodia formation.
Specifications
Cdc42EP3 | |
Polyclonal | |
Unconjugated | |
CDC42EP3 | |
3200001F04Rik; AA986861; Binder of Rho GTPases 2; BORG2; CDC42 effector protein (Rho GTPase binding) 3; CDC42 effector protein 3; Cdc42ep3; CEP3; CRIB-containing BORG2 protein; MSE55-related Cdc42-binding protein; MSE55-related protein; UB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKI2 | |
CDC42EP3 | |
Recombinant protein corresponding to Human CDC42EP3. Recombinant protein control fragment (Product #RP-108461). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction