Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42EP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3136 Supplier Bioss Supplier No. BS13759R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. This protein can interact with CDC42, as well as with the ras homolog gene family, member Q (ARHQ/TC10). Expression of this protein in fibroblasts has been shown to induce pseudopodia formation.

Specifications

Antigen CDC42EP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDC42EP3
Gene Accession No. Q9UKI2
Gene Alias 3200001F04Rik; AA986861; Binder of Rho GTPases 2; Borg2; CDC42 effector protein (Rho GTPase binding) 3; Cdc42 effector protein 3; CDC42EP3; Cep3; CRIB-containing BORG2 protein; MSE55-related Cdc42-binding protein; MSE55-related protein; UB1
Gene Symbols CDC42EP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDC42EP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10602
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.