Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc42EP3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572573
Description
This gene encodes a member of a small family of guanosine triphosphate (GTP) metabolizing proteins that contain a CRIB (Cdc42, Rac interactive binding) domain. Members of this family of proteins act as effectors of CDC42 function. The encoded protein is involved in actin cytoskeleton re-organization during cell shape changes, including pseudopodia formation. A pseudogene of this gene is found on chromosome 19. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2012].
Specifications
Cdc42EP3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UKI2 | |
CDC42EP3 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CDC42EP3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CDC42EP3 | |
Cdc42 effector protein 3; Binder of Rho GTPases 2; MSE55-related Cdc42-binding protein; CDC42EP3; BORG2; CEP3 | |
Rabbit | |
Protein A | |
RUO | |
10602 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction