Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc42EP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554769 Supplier Invitrogen™ Supplier No. PA554769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHTMHVGRAG DAFGDTSFLN SKAGEPDGES LDEQPSSSSS KRSLLSRKFR GSKRSQSVTR GEREQRDMLG SLRDSALFVK NAMSLPQLNE KEAAEKGTSK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

The product of this gene is a member of the CDC42-binding protein family. Members of this family interact with Rho family GTPases and regulate the organization of the actin cytoskeleton. This protein has been shown to bind both CDC42 and TC10 GTPases in a GTP-dependent manner. When overexpressed in fibroblasts, this protein was able to induce pseudopodia formation, which suggested a role in inducing actin filament assembly and cell shape control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdc42ep4
Gene Accession No. Q9H3Q1
Gene Alias 1500041M20Rik; binder of Rho GTPase 4; binder of Rho GTPases 4; BORG4; CDC42 effector protein (Rho GTPase binding) 4; CDC42 effector protein 4; CDC42EP4; Cep4; KAIA1777; MGC17125; MGC3740; Unknown (protein for MGC:138126)
Gene Symbols Cdc42ep4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cdc42EP4. Recombinant protein control fragment (Product #RP-93478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.