Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc42EP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570044 Supplier Thermo Scientific Supplier No. PA570044
Add to Cart
Add to Cart

Cdc42EP4 Polyclonal Antibody

Cdc42EP4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Cdc42ep4
Gene Accession No. 0
Gene Alias 1500041M20Rik; binder of Rho GTPase 4; binder of Rho GTPases 4; BORG4; CDC42 effector protein (Rho GTPase binding) 4; CDC42 effector protein 4; CDC42EP4; Cep4; KAIA1777; MGC17125; MGC3740; Unknown (protein for MGC:138126)
Gene Symbols Cdc42ep4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human Cdc42EP4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303653
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.