Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc42EP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106636 Supplier Invitrogen™ Supplier No. PA5106636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BORG4.

The product of this gene is a member of the CDC42-binding protein family. Members of this family interact with Rho family GTPases and regulate the organization of the actin cytoskeleton. This protein has been shown to bind both CDC42 and TC10 GTPases in a GTP-dependent manner. When overexpressed in fibroblasts, this protein was able to induce pseudopodia formation, which suggested a role in inducing actin filament assembly and cell shape control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdc42ep4
Gene Accession No. Q9H3Q1, Q9JM96
Gene Alias 1500041M20Rik; binder of Rho GTPase 4; binder of Rho GTPases 4; BORG4; CDC42 effector protein (Rho GTPase binding) 4; CDC42 effector protein 4; CDC42EP4; Cep4; KAIA1777; MGC17125; MGC3740; Unknown (protein for MGC:138126)
Gene Symbols Cdc42ep4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDC42EP4(Accession Q9H3Q1), corresponding to amino acid residues A203-P253.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23580, 303653, 56699
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.