Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc42EP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110907
Description
Immunogen sequence: PVKKANDGEG GDEEAGTEEA VPRRNGAAGP HSPDPLLDEQ AFGDLTDLPV VPKATYGLKH AESIMSFHID LGPSMLGDVL SIMDKEEWDP EEGEGGYHGD EGA.
The product of this gene is a member of the CDC42-binding protein family. Members of this family interact with Rho family GTPases and regulate the organization of the actin cytoskeleton. This protein has been shown to bind both CDC42 and TC10 GTPases in a GTP-dependent manner. When overexpressed in fibroblasts, this protein was able to induce pseudopodia formation, which suggested a role in inducing actin filament assembly and cell shape control.
Specifications
| Cdc42EP4 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3Q1 | |
| Cdc42ep4 | |
| Recombinant Protein corresonding to Human Cdc42EP4. Recombinant protein control fragment (Product #RP-93479). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Cdc42ep4 | |
| 1500041M20Rik; binder of Rho GTPase 4; binder of Rho GTPases 4; BORG4; CDC42 effector protein (Rho GTPase binding) 4; CDC42 effector protein 4; CDC42EP4; Cep4; KAIA1777; MGC17125; MGC3740; Unknown (protein for MGC:138126) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23580 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction