Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc42EP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560367 Supplier Thermo Scientific Supplier No. PA560367
Add to Cart
Add to Cart

Cdc42EP5 Polyclonal Antibody

Cdc42EP5 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC42EP5
Gene Accession No. Q6NZY7
Gene Alias 1700027J19Rik; 2010007O02Rik; binder of Rho GTPase 3; binder of Rho GTPases 3; BORG3; C85526; CDC42 effector protein (Rho GTPase binding) 5; CDC42 effector protein 5; CDC42EP5; Cep5; CRIB-containing BORG3 protein
Gene Symbols CDC42EP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cdc42EP5. Recombinant protein control fragment (Product #RP-99145).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.