Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc42EP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106637
Description
Antibody detects endogenous levels of total BORG3.
Cell division control protein 42 (CDC42), a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg (binder of Rho GTPases) family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to CDC42 and regulate its function negatively. The encoded protein may inhibit c-Jun N-terminal kinase (JNK) independently of CDC42 binding. The protein may also play a role in septin organization and inducing pseudopodia formation in fibroblasts [provided by RefSeq, Jul 2013].
Specifications
| Cdc42EP5 | |
| Polyclonal | |
| Unconjugated | |
| CDC42EP5 | |
| 1700027J19Rik; 2010007O02Rik; binder of Rho GTPase 3; binder of Rho GTPases 3; BORG3; C85526; CDC42 effector protein (Rho GTPase binding) 5; CDC42 effector protein 5; CDC42EP5; Cep5; CRIB-containing BORG3 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 148170, 58804 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NZY7, Q9Z0X0 | |
| CDC42EP5 | |
| A synthesized peptide derived from human CDC42EP5(Accession Q6NZY7), corresponding to amino acid residues M1-R38. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction