Learn More
Cdc42GAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Cdc42GAP |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2400
![]() |
Bethyl Laboratories, Inc
IHC00610T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2399
![]() |
Bethyl Laboratories, Inc
IHC00610 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Guinea pig_10141, Orangutan, Monkey, Chimpanzee, Northern white-cheeked gibbo
GTPase activator for the Rho, Rac and Cdc42 proteins, converting them to the putatively inactive GDP-bound state. Cdc42 seems to be the preferred substrate.Specifications
Cdc42GAP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP | |
Between 1 and 50 | |
Primary | |
4° C | |
ARHGAP1 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q07960, Q5FWK3 | |
228359, 392 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.