Learn More
Cdc42GAP Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301853A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GTPase activator for the Rho, Rac and Cdc42 proteins, converting them to the putatively inactive GDP-bound state. Cdc42 seems to be the preferred substrate.Specifications
Cdc42GAP | |
Polyclonal | |
Unconjugated | |
ARHGAP1 | |
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228359, 392 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q07960, Q5FWK3 | |
ARHGAP1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.