Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC42SE2 Antibody (OTI1B12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272070F
Description
CDC42SE2 Monoclonal specifically detects CDC42SE2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
CDC42SE2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CDC42SE2 | |
Full length recombinant protein of human CDC42SE2 (NP_064625) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI1B12 | |
Immunohistochemistry | |
CDC42 small effector 2, FLJ21967, non-kinase Cdc42 effector protein SPEC2, Small effector of CDC42 protein 2, SPEC2CDC42 small effector protein 2 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
56990.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction