Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC42SE2 Antibody (OTI1B12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246191
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CDC42SE2 Monoclonal specifically detects CDC42SE2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CDC42SE2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
CDC42 small effector 2, FLJ21967, non-kinase Cdc42 effector protein SPEC2, Small effector of CDC42 protein 2, SPEC2CDC42 small effector protein 2 | |
Mouse | |
9 kDa | |
0.1 mL | |
Protein Kinase | |
56990 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1B12 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CDC42SE2 | |
Full length human recombinant protein of human CDC42SE2 (NP_064625) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction