Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDC42SE2 Monoclonal Antibody (OTI11F7)
GREENER_CHOICE

Catalog No. PIMA526954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526954 Supplier Invitrogen™ Supplier No. MA526954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDC42SE2 Monoclonal Antibody for Western Blot, IHC (P)

Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42SE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI11F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDC42SE2
Gene Accession No. Q9NRR3
Gene Alias 2810404F18Rik; AA408783; AA536669; CDC42 small effector 2; CDC42 small effector protein 2; Cdc42se2; non-kinase Cdc42 effector protein SPEC2; RGD1563924; small effector of CDC42 protein 2; SPEC2
Gene Symbols CDC42SE2
Host Species Mouse
Immunogen Full length human recombinant protein of CDC42SE2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56990
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.