Learn More
CDC42SE2 Mouse anti-Human, Clone: OTI1B2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808181

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.Specifications
CDC42SE2 | |
Monoclonal | |
Unconjugated | |
CDC42SE2 | |
SPEC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
56990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NRR3 | |
CDC42SE2 | |
Full length human recombit protein of human CDC42SE2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.