Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc45L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529163
Description
Recommended positive controls: Jurkat, Raji, NCI-H929, A549, HeLa, AGS. Predicted reactivity: Mouse (93%), Rat (92%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene was identified by its strong similarity with Saccharomyces cerevisiae Cdc45, an essential protein required to the initiation of DNA replication. Cdc45 is a member of the highly conserved multiprotein complex including Cdc6/Cdc18, the minichromosome maintenance proteins (MCMs) and DNA polymerase, which is important for early steps of DNA replication in eukaryotes. This protein has been shown to interact with MCM7 and DNA polymerase alpha. Studies of the similar gene in Xenopus suggested that this protein play a pivotal role in the loading of DNA polymerase alpha onto chromatin. Multiple polyadenylation sites of this gene are reported.
Specifications
| Cdc45L | |
| Polyclonal | |
| Unconjugated | |
| Cdc45 | |
| CDC 45; CDC 45L; Cdc45; CDC45 cell division cycle 45 homolog; CDC45 like; CDC45L; CDC45L2; CDC45-related protein; Cell division control protein 45 homolog; cell division cycle 45; cell division cycle 45 homolog; cell division cycle 45-like 2; human CDC45; PORC PI 1; PORC-PI-1; UNQ374/PRO710 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12544, 8318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75419, Q9Z1X9 | |
| Cdc45 | |
| Recombinant protein encompassing a sequence within the center region of human CDC45L. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction