Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc5L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552823
Description
Immunogen sequence: GAEGLTPRSG TTPKPVINST PGRTPLRDKL NINPEDGMAD YSDPSYVKQM ERESREHLRL GLLGLPAPKN DFEIVLPENA EKELEEREID DTYIEDAADV DARKQAIRDA ERVKEMKRMH KAVQKDLPRP SEVNETILR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing.
Specifications
Cdc5L | |
Polyclonal | |
Unconjugated | |
CDC5L | |
1200002I02Rik; AA408004; CDC5; CDC5 cell division cycle 5-like; CDC5 cell division cycle 5-like (S. pombe); CDC5L; CDC5-LIKE; Cdc5-like protein; Cdc5-related protein; CEF1; cell division cycle 5 like; cell division cycle 5-like; cell division cycle 5-like (S. pombe); cell division cycle 5-like protein; cell division cycle 5-related protein; dJ319D22.1; dJ319D22.1 (CDC5-like protein); hCDC5; KIAA0432; Pcdc5rp; Pombe cdc5-related protein; Unknown (protein for MGC:128482); zgc:55853 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
988 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99459 | |
CDC5L | |
Recombinant protein corresponding to Human Cdc5L. Recombinant protein control fragment (Product #RP-102482). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction