Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595132
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595132 Supplier Invitrogen™ Supplier No. PA595132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human THP-1 whole cell, rat C6 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc5L
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene CDC5L
Gene Accession No. O08837, Q99459
Gene Alias 1200002I02Rik; AA408004; CDC5; CDC5 cell division cycle 5-like; CDC5 cell division cycle 5-like (S. pombe); CDC5L; CDC5-LIKE; Cdc5-like protein; Cdc5-related protein; CEF1; cell division cycle 5 like; cell division cycle 5-like; cell division cycle 5-like (S. pombe); cell division cycle 5-like protein; cell division cycle 5-related protein; dJ319D22.1; dJ319D22.1 (CDC5-like protein); hCDC5; KIAA0432; Pcdc5rp; Pombe cdc5-related protein; Unknown (protein for MGC:128482); zgc:55853
Gene Symbols CDC5L
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human CDC5L (782-802aa KELQHRYADLLLEKETLKSKF).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85434, 988
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.