Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc5L Recombinant Rabbit Monoclonal Antibody (22H12L1)

Catalog No. 702831
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-283-1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-283-1 Supplier Invitrogen™ Supplier No. 702831
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Rabbit, Cat, Horse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc5L
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 22H12L1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene CDC5L
Gene Accession No. Q6A068, Q99459
Gene Alias 1200002I02Rik; AA408004; CDC5; CDC5 cell division cycle 5-like; CDC5 cell division cycle 5-like (S. pombe); CDC5L; CDC5-LIKE; Cdc5-like protein; Cdc5-related protein; CEF1; cell division cycle 5 like; cell division cycle 5-like; cell division cycle 5-like (S. pombe); cell division cycle 5-like protein; cell division cycle 5-related protein; dJ319D22.1; dJ319D22.1 (CDC5-like protein); hCDC5; KIAA0432; Pcdc5rp; Pombe cdc5-related protein; Unknown (protein for MGC:128482); zgc:55853
Gene Symbols CDC5L
Host Species Rabbit
Immunogen Protein corresponding to human CDC5L [aa1-aa200].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71702, 988
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.