Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc6 Antibody (Cdc6 9H8/5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002684AF647
Description
Cdc6 Monoclonal specifically detects Cdc6 in Yeast samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cdc6 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDC6 | |
Cdc6 from S. cerevisiae. | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
990 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
Cdc6 9H8/5 | |
Western Blot, Immunohistochemistry | |
CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction