Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc6 Antibody (OTI1A4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270383B
Description
Cdc6 Monoclonal specifically detects Cdc6 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cdc6 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CDC6 | |
Full length recombinant protein of human Cdc6 (NP_001245) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A4 | |
Western Blot, Immunohistochemistry | |
CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
990.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction