Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577901
Description
Positive Control: Jurkat, HeLa, HeLa (10 mM Hydroxyurea treatment for 18 hr), Neuro2A, GL261, C8D30, BCL-1, Raw264.7, C2C12, PC-12, Rat2 Predicted Reactivity: Bovine (89%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.
Specifications
Cdc6 | |
Polyclonal | |
Unconjugated | |
CDC6 | |
CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23834, 360621, 990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.36 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O89033, Q99741 | |
CDC6 | |
Recombinant protein encompassing a sequence within the center region of human CDC6. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction