Learn More
Promega CDC7/DBF4 Kinase Enzyme System, 1mg
Catalog No. PRV6577 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6577 1 mg
1 options
Catalog No. PRV6577 Supplier Promega Supplier No. V6577
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CDC7 and DBF4 proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile CDC7/DBF4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C