Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC7 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562754
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2754 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2754 Supplier Bethyl Laboratories, Inc Supplier No. A302504AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CDC7L1, CDC7 type protein kinase, regulates firing of the origins of initiation in DNA replication. CDC7L1 forms a complex with a Dbf4-related regulatory subunit to generate an active kinase. Mini-chromosome maintece protein (MCM) is the major physiological target of Cdc7L1. At least three mRNA variants have been reported. Overexpression of the CDC7L1 protein may be associated with neoplastic transformation for some tumors. CDC7L1 has been documented in a wide range of tissues. One variant is expressed ubiquitously in human tissues while at least one other has been shown to be testis-specific. ESTs have been isolated from several tissue libraries, including normal embryo, gallbladder, lymph node and thymus and cancerous duodenum and embryo.

Specifications

Antigen CDC7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CDC7
Gene Accession No. O00311
Gene Alias CDC7L1, HsCDC7, Hsk1, MGC117361, MGC126237, MGC126238, huCDC7
Gene Symbols CDC7
Host Species Rabbit
Immunogen Between 524 and 574
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8317
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.