Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986365
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6365 Supplier Bioss Supplier No. BS5148R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a cell division cycle protein with kinase activity that is critical for the G1/S transition. The yeast homolog is also essential for initiation of DNA replication as cell division occurs. Overexpression of this gene product may be associated with neoplastic transformation for some tumors. Multiple alternatively spliced transcript variants that encode the same protein have been detected.

Specifications

Antigen Cdc7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDC7
Gene Accession No. O00311
Gene Alias AI597260; Cdc7; CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1; Cdc7 Kinase; Cdc7l1; CDC7-like 1; CDC7-related kinase; cell division cycle 7; cell division cycle 7 (S. cerevisiae); cell division cycle 7 homolog; cell division cycle 7-like 1; cell division cycle 7-like protein 1; Cell division cycle 7-related protein kinase; HsCDC7; Hsk1; huCDC7; MGC117361; MGC126237; MGC126238; MGC91067; muCdc7
Gene Symbols CDC7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Cdc7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.