Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC7 Rabbit anti-Human, Unconjugated, Clone: BGF-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19870
Description
CDC7 Monoclonal antibody specifically detects CDC7 in Human samples. It is validated for WB-Ce, Flow Cyt.Specifications
CDC7 | |
Monoclonal | |
Unconjugated | |
CDC7 | |
Rabbit | |
100 μg | |
8317 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Flow Cytometry | |
BGF-3 | |
cell division cycle 7 homolog (S. cerevisiae) | |
CDC7L1, HsCDC7, Hsk1, MGC117361, MGC126237, MGC126238, huCDC7 | |
A synthetic peptide corresponding to human CDC7 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction