Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC73/HRPT2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200184V
Description
CDC73/HRPT2 Polyclonal antibody specifically detects CDC73/HRPT2 in Human, Mouse samples. It is validated for Western BlotSpecifications
CDC73/HRPT2 | |
Polyclonal | |
Western Blot | |
C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human Parafibromin using the numbering given in TrEMBL entry Q6P1J9 (GeneID 79577). | |
0.1 mL | |
Cancer, Tumor Suppressors | |
79577 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction