Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC73/HRPT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200184
Description
CDC73/HRPT2 Polyclonal specifically detects CDC73/HRPT2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| CDC73/HRPT2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB200-184 is specific for human Parafibromin protein. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:5000-1:20000, Immunoprecipitation 1-4 ug/mg lysate | |
| Q6P1J9 | |
| CDC73 | |
| The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human Parafibromin using the numbering given in TrEMBL entry Q6P1J9 (GeneID 79577). | |
| 100 μL | |
| Cancer, Tumor Suppressors | |
| 79577 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction