Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC73/HRPT2 Antibody (SR2070), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32263750UL
Description
CDC73/HRPT2 Monoclonal antibody specifically detects CDC73/HRPT2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CDC73/HRPT2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2070 | |
Western Blot 1:500-1:2000 | |
C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
A synthesized peptide derived from human CDC73/HRPT2 (Uniprot #: Q6P1J9) | |
50 μg | |
Cancer, Tumor Suppressors | |
79577 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction