Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc73 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526189

Catalog No. PIPA526189


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

Cdc73 is a tumor suppressor probably involved in transcriptional and post-transcriptional control pathways. Cdc73 is part of the PAF1 complex, a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9. PAF1 was first purified from yeast in association with RNA polymerase II and is believed to function in transcription elongation. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. In addition to coordinating events during transcription (initiation, promoter clearance, and elongation), hPAF also coordinates events in RNA quality control. Cdc73 may also be involved in cell cycle progression through the regulation of cyclin D1/PRAD1 expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdc73
Polyclonal
Unconjugated
CDC73
8430414L16Rik; BC027756; C130030P16Rik; C1orf28; C81219; Cdc73; cell division cycle 73; cell division cycle 73 Paf1/RNA polymerase II complex component-like protein; cell division cycle 73, Paf1/RNA polymerase II complex component; cell division cycle 73, Paf1/RNA polymerase II complex component, homolog; cell division cycle protein 73 homolog; FIHP; HPTJT; HRPT1; HRPT2; hyperparathyroidism 2 (with jaw tumor); hyperparathyroidism 2 homolog; hyperparathyroidism 2 protein; Hyperparathyroidism 2 protein homolog; HYX; Paf1/RNA polymerase II complex component; parafibromin; Vcell division cycle 73, Paf1/RNA polymerase II complex component, homolog
Rabbit
Protein A, Antigen affinity chromatography
RUO
79577
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6P1J9
CDC73
KLH conjugated synthetic peptide between 132-161 amino acids from the central region of human CDC73.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.