Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556322 Supplier Invitrogen™ Supplier No. PA556322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKCERKDDFL GAAEGKLQCN RLMPNSQKDC HCLGDVLIEN TKESKSQSED LGRKPMESSS VVSCRDRKDR RRSMCYSDGR SLHLEKNGNH TPSS Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 46%.

Cell division cycle associated 2 (CDCA2, Repo-Man) is a cell-cycle protein that recruits protein phosphatase 1 (PP1) to mitotic chromatin at anaphase onset, which is essential for cell proliferation. Carboxy-terminal phosphorylation of CDCA2 at Ser893 by Aurora B inhibits the protein and leads to diffuse localization during prometaphase and metaphase. Dephosphorylation of CDCA2 by PP2A is necessary for CDCA2/PP1 complex reformation. The CDCA2/PP1 complex is required for chromatin binding and dephosphorylation of histone H3 at Thr3, Ser10, and Ser28. The CDCA2/PP1 complex is also involved in nuclear envelope reformation during mitotic exit for proper progression through the M/G1 transition. The interaction of CDCA2 with importin beta and Nup153, which is required for nuclear envelope formation, is negatively regulated by CDK phosphorylation of the amino-terminal domain of CDCA2. CDCA2 may play a role in DNA repair as the release of CDCA2 from chromatin at sites of DNA damage promotes the activation of DNA damage response. These results imply that the CDCA2/PP1 complex may play a part in cancer progression. Research studies indicate that CDCA2 may serve as a prognostic marker, as increased CDCA2 expression is seen in a number of cancers, including melanoma, neuroblastoma tumors, squamous cell carcinoma, and synovial sarcomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDCA2
Gene Accession No. Q69YH5
Gene Alias 2610311M19Rik; AI586158; CDCA2; cell division cycle associated 2; cell division cycle-associated protein 2; FLJ25804; MGC129906; MGC129907; PPP1R81; protein phosphatase 1, regulatory subunit 81; recruits PP1 onto mitotic chromatin at anaphase protein; Repo-Man
Gene Symbols CDCA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDCA2. Recombinant protein control fragment (Product #RP-93942).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.