Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501731729 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1729 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1729 Supplier Proteintech Group Inc Supplier No. 177011AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cell division cycle associated 2 (CDCA2, Repo-Man) is a cell-cycle protein that recruits protein phosphatase 1 (PP1) to mitotic chromatin at anaphase onset, which is essential for cell proliferation. Carboxy-terminal phosphorylation of CDCA2 at Ser893 by Aurora B inhibits the protein and leads to diffuse localization during prometaphase and metaphase. Dephosphorylation of CDCA2 by PP2A is necessary for CDCA2/PP1 complex reformation. The CDCA2/PP1 complex is required for chromatin binding and dephosphorylation of histone H3 at Thr3, Ser10, and Ser28. The CDCA2/PP1 complex is also involved in nuclear envelope reformation during mitotic exit for proper progression through the M/G1 transition. The interaction of CDCA2 with importin beta and Nup153, which is required for nuclear envelope formation, is negatively regulated by CDK phosphorylation of the amino-terminal domain of CDCA2. CDCA2 may play a role in DNA repair as the release of CDCA2 from chromatin at sites of DNA damage promotes the activation of DNA damage response. These results imply that the CDCA2/PP1 complex may play a part in cancer progression. Research studies indicate that CDCA2 may serve as a prognostic marker, as increased CDCA2 expression is seen in a number of cancers, including melanoma, neuroblastoma tumors, squamous cell carcinoma, and synovial sarcomas.

Specifications

Antigen CDCA2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDCA2
Gene Accession No. Q69YH5
Gene Alias CDCA2, Repo Man
Gene Symbols CDCA2
Host Species Rabbit
Immunogen CDCA2 Fusion Protein Ag11967
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.